Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100700.1 | NP_001094170.1 | 3 | 956 | ||
GenBank mRNA | AJ560715.1 | - | 1 | 94 | ||
BC161900.1 | - | 3 | 956 | 8367121 |
Set Membership: |
G-protein coupled receptor |