Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107749.1 | NP_001101219.1 | 4 | 396 | ||
XM_006234569.2 | XP_006234631.1 | 4 | 624 | |||
XM_006234570.2 | XP_006234632.1 | 4 | 585 | |||
XM_006234571.2 | XP_006234633.1 | 3 | 251 | |||
XM_006234572.2 | XP_006234634.1 | 4 | 434 | |||
GenBank mRNA | BC167758.1 | - | 4 | 534 | 7123311 |