Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014053.1 | NP_001014075.1 | 2 | 957 | ||
GenBank mRNA | BC061729.1 | - | 2 | 957 | 5598709 |