Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229179.2 | XP_006229241.1 | 4 | 534 | ||
XM_006229180.2 | XP_006229242.1 | 4 | 534 | |||
XM_006229182.2 | XP_006229244.1 | 4 | 534 | |||
XM_006229183.2 | XP_006229245.1 | 3 | 415 | |||
XM_008759449.1 | XP_008757671.1 | 4 | 534 | |||
XM_008759450.1 | XP_008757672.1 | 4 | 591 | |||
XM_341847.7 | XP_341848.3 | 4 | 534 |