Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108650.2 | NP_001102120.1 | 3, 4 | 370 | ||
XM_008763394.1 | XP_008761616.1 | 3, 4 | 1101 | |||
XM_008763395.1 | XP_008761617.1 | 3, 4 | 1101 | |||
GenBank mRNA | BC160872.1 | - | 3, 4 | 370 | 7365768 | |
FQ211769.1 | - | 3, 4 | 465 |