Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024757.1 | NP_001019928.1 | 4 | 364 | ||
XM_006237930.2 | XP_006237992.1 | 4 | 279 | |||
XM_006237931.2 | XP_006237993.1 | 3 | 706 | |||
XM_008763590.1 | XP_008761812.1 | 5 | 577 | |||
GenBank mRNA | BC097386.1 | - | 4 | 364 | 7446512 |