Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019291.1 | NP_062164.1 | 6, 7 | 668 | ||
GenBank mRNA | BC065577.1 | - | 6, 7 | 701 | 5623601 | |
FQ212832.1 | - | 6, 7 | 720 | |||
FQ213417.1 | - | 6, 7 | 720 | |||
FQ214016.1 | - | 6, 7 | 720 | |||
FQ225794.1 | - | 6, 7 | 718 | |||
FQ225882.1 | - | 6, 7 | 718 | |||
FQ225962.1 | - | 6, 7 | 718 | |||
FQ226264.1 | - | 6, 7 | 720 | |||
FQ231140.1 | - | 6, 7 | 718 | |||
FQ232067.1 | - | 6, 7 | 718 | |||
FQ233035.1 | - | 6, 7 | 719 | |||
FQ233398.1 | - | 6, 7 | 718 | |||
FQ234177.1 | - | 6, 7 | 717 | |||
FQ234222.1 | - | 6, 7 | 718 | |||
FQ234668.1 | - | 6, 7 | 720 | |||
X58294.1 | - | 6, 7 | 668 |