Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020471.1 | NP_065204.1 | 2 | 177 | ||
XM_006243089.1 | XP_006243151.1 | 2 | 336 | |||
GenBank mRNA | L35571.1 | - | 2 | 177 |