Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035236.1 | NP_001030313.1 | 4 | 552 | ||
NM_031676.2 | NP_113864.2 | 3 | 500 | |||
GenBank mRNA | AF459788.1 | - | 4 | 696 | ||
FQ212703.1 | - | 3 | 494 | |||
FQ214377.1 | - | 3 | 493 | |||
FQ214483.1 | - | 3 | 494 | |||
FQ214564.1 | - | 3 | 494 | |||
M84725.1 | - | 3 | 474 |
Gene Symbol: LOC103693564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768676.1 | XP_008766898.1 | 4 | 769 |