Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233339.2 | XP_006233401.1 | 3 | 1175 | ||
XM_006233341.2 | XP_006233403.1 | 2 | 1090 | |||
XM_008761529.1 | XP_008759751.1 | 2 | 1251 | |||
XM_008761530.1 | XP_008759752.1 | 2 | 1251 | |||
XM_008761531.1 | XP_008759753.1 | 2 | 1251 | |||
GenBank mRNA | AF272158.1 | - | 1 | 314 |