Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101021.1 | NP_001094491.1 | 884 | |||
GenBank mRNA | DQ256461.1 | - | 884 |