Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100125367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103357.1 | NP_001096827.1 | 2 | 489 | ||
XM_008769815.1 | XP_008768037.1 | 2 | 411 | |||
GenBank mRNA | BC085897.1 | - | 2 | 489 | 7193261 |