Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103361.1 | NP_001096831.1 | 1 | 561 | ||
XM_008765081.1 | XP_008763303.1 | 1 | 724 | |||
XM_008765082.1 | XP_008763304.1 | 1 | 1880 | |||
GenBank mRNA | BC093394.1 | - | 1 | 561 | 7388692 |