Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100151767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127503.1 | NP_001120975.1 | 5 | 364 | ||
XM_006243024.2 | XP_006243086.1 | 5 | 377 | |||
XM_006243025.2 | XP_006243087.1 | 5 | 896 | |||
XM_006243026.2 | XP_006243088.1 | 5 | 896 | |||
GenBank mRNA | BC161965.1 | - | 5 | 364 | 9028305 |