Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236798.2 | XP_006236860.1 | 3 | 850 | ||
GenBank mRNA | AF187814.1 | - | 1 | 59 | ||
FQ213004.1 | - | 2 | 221 |