Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080777.2 | NP_542955.2 | 2 | 211 | ||
XM_006253594.2 | XP_006253656.1 | 3 | 412 | |||
GenBank mRNA | BC087579.1 | - | 2 | 211 | 7314241 | |
D17764.1 | - | 2 | 67 |