Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147135.2 | NP_671476.1 | 3 | 863 | ||
XM_008759853.1 | XP_008758075.1 | 3 | 870 | |||
XM_008759854.1 | XP_008758076.1 | 4 | 1198 | |||
GenBank mRNA | AB010154.1 | - | 3 | 575 | ||
BC131843.1 | - | 3 | 863 | 8363395 |
Gene Symbol: LOC103690016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758862.1 | XP_008757084.1 | 3 | 911 | ||
XM_008758863.1 | XP_008757085.1 | 3 | 868 | |||
XM_008758864.1 | XP_008757086.1 | 4 | 1172 |