Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053634.1 | NP_446086.1 | 9 | 951 | ||
GenBank mRNA | AB036792.1 | - | 9 | 951 | ||
BC088123.1 | - | 9 | 943 | 7376834 | ||
FQ220557.1 | - | 9 | 954 | |||
FQ233041.1 | - | 9 | 954 |