Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053693.1 | NP_446145.1 | 8 | 828 | ||
XM_006235993.1 | XP_006236055.1 | 7 | 680 | |||
GenBank mRNA | BC081880.1 | - | 8 | 828 | 7108159 |