Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053713.1 | NP_446165.1 | 3 | 568 | ||
GenBank mRNA | AF390546.1 | - | 3 | 568 | ||
BC085720.1 | - | 3 | 961 | 7190219 |