Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053732.1 | NP_446184.1 | 1, 2 | 1468 | ||
XM_008767531.1 | XP_008765753.1 | 1, 2 | 3023 | |||
GenBank mRNA | AY028418.1 | - | 1, 2 | 926 |