Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053756.1 | NP_446208.1 | 4 | 514 | ||
XM_006234393.2 | XP_006234455.1 | 5 | 543 | |||
XM_006234394.2 | XP_006234456.1 | 5 | 523 | |||
GenBank mRNA | AF315374.1 | - | 4 | 514 | ||
BC099786.1 | - | 4 | 426 | 7934752 | ||
FQ217141.1 | - | 5 | 481 | |||
FQ223732.1 | - | 5 | 491 | |||
FQ224352.1 | - | 4 | 449 |