Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053776.2 | NP_446228.2 | 13 | 1443 | ||
XM_008762628.1 | XP_008760850.1 | 14 | 1752 | |||
XM_008762629.1 | XP_008760851.1 | 11 | 1005 | |||
GenBank mRNA | AF118853.1 | - | 13 | 1416 | ||
AY322161.1 | - | 13 | 1416 | |||
AY322162.1 | - | 12 | 1492 | |||
AY322163.1 | - | 13 | 1327 | |||
AY322164.1 | - | 6 | 492 | |||
BC088838.1 | - | 13 | 1443 | 7132881 | ||
FQ227102.1 | - | 5 | 453 |