Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053825.2 | NP_446277.1 | 5 | 613 | ||
XM_006254229.2 | XP_006254291.1 | 5 | 667 | |||
XM_006254230.2 | XP_006254292.1 | 5 | 668 | |||
XM_006254231.2 | XP_006254293.1 | 5 | 579 | |||
GenBank mRNA | BC059158.1 | - | 5 | 617 | 6920493 | |
FQ212577.1 | - | 5 | 601 | |||
FQ212651.1 | - | 5 | 611 | |||
FQ212813.1 | - | 5 | 611 | |||
FQ213008.1 | - | 5 | 611 | |||
FQ213785.1 | - | 5 | 613 | |||
FQ213852.1 | - | 5 | 613 | |||
FQ213883.1 | - | 5 | 607 | |||
FQ214745.1 | - | 5 | 591 | |||
FQ214863.1 | - | 5 | 667 | |||
FQ214998.1 | - | 5 | 611 | |||
FQ215286.1 | - | 5 | 586 | |||
FQ215351.1 | - | 5 | 611 | |||
FQ215426.1 | - | 5 | 610 | |||
FQ215847.1 | - | 5 | 611 | |||
FQ216198.1 | - | 5 | 607 | |||
FQ216285.1 | - | 5 | 601 | |||
FQ216490.1 | - | 5 | 611 | |||
FQ216539.1 | - | 5 | 595 | |||
FQ216647.1 | - | 5 | 613 | |||
FQ218995.1 | - | 5 | 613 | |||
FQ219484.1 | - | 5 | 611 | |||
FQ219563.1 | - | 5 | 611 | |||
FQ219689.1 | - | 5 | 613 | |||
FQ220076.1 | - | 5 | 611 | |||
FQ220217.1 | - | 5 | 613 | |||
FQ220355.1 | - | 5 | 611 | |||
FQ224939.1 | - | 5 | 611 | |||
FQ229987.1 | - | 5 | 634 | |||
FQ234981.1 | - | 5 | 611 | |||
L19927.1 | - | 3 | 491 |