Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053849.1 | NP_446301.1 | 10 | 2159 | ||
GenBank mRNA | BC061535.1 | - | 10 | 1973 | 6922170 | |
M86870.1 | - | 10 | 2159 |