Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760849.1 | XP_008759071.1 | 5 | 1870 | ||
XM_574892.6 | XP_574892.1 | 6 | 1341 | |||
GenBank mRNA | U31668.1 | - | 6 | 690 |