Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053946.1 | NP_446398.1 | 4, 5 | 572 | ||
GenBank mRNA | AF008554.2 | - | 4, 5 | 572 |