Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3f3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053985.2 | NP_446437.1 | 2 | 168 | ||
GenBank mRNA | BC063159.1 | - | 2 | 168 | 6921452 | |
BC078759.1 | - | 2 | 137 | 7112262 | ||
FQ215473.1 | - | 2 | 161 | |||
FQ225133.1 | - | 2 | 163 | |||
FQ225528.1 | - | 2 | 161 | |||
FQ226787.1 | - | 2 | 161 | |||
FQ227040.1 | - | 2 | 163 | |||
FQ227957.1 | - | 2 | 163 | |||
FQ228569.1 | - | 2 | 163 | |||
FQ232451.1 | - | 2 | 97 | |||
FQ232671.1 | - | 2 | 161 | |||
FQ232852.1 | - | 2 | 163 | |||
FQ234172.1 | - | 2 | 163 | |||
FQ234408.1 | - | 2 | 161 | |||
FQ235236.1 | - | 2 | 166 | |||
X73683.1 | - | 2 | 159 |