Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053991.1 | NP_446443.1 | 6 | 638 | ||
XM_006227827.2 | XP_006227889.1 | 7 | 1080 | |||
XM_006227828.2 | XP_006227890.1 | 7 | 1083 | |||
GenBank mRNA | AB089807.1 | - | 6 | 560 | ||
AB089808.1 | - | 5 | 795 | |||
AB089809.1 | - | 6 | 655 | |||
BC158798.1 | - | 6 | 635 | 7316540 | ||
X02341.1 | - | 5 | 473 |