Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301286.1 | NP_001288215.1 | 1 | 54 | ||
NM_057208.2 | NP_476556.2 | 1 | 54 | |||
NM_173111.1 | NP_775134.1 | 1 | 96 | |||
XM_006232580.2 | XP_006232642.1 | 1 | 154 | |||
XM_006232581.2 | XP_006232643.1 | 1 | 155 | |||
XM_006232583.2 | XP_006232645.1 | 1 | 158 | |||
XM_006232584.2 | XP_006232646.1 | 1 | 156 | |||
XM_008761124.1 | XP_008759346.1 | 1 | 154 | |||
XM_008761126.1 | XP_008759348.1 | 1 | 155 | |||
GenBank mRNA | FQ228755.1 | - | 1 | 104 | ||
L24775.1 | - | 1 | 54 | |||
L24776.1 | - | 54 | ||||
L24777.1 | - | 1 | 54 | |||
S82383.1 | - | 1 | 73 | |||
X72859.1 | - | 1 | 96 |