Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139036.1 | NP_620605.1 | 4 | 1159 | ||
GenBank mRNA | L35572.1 | - | 4 | 1159 |