Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080409.1 | NP_536334.1 | 17 | 2362 | ||
XM_008769527.1 | XP_008767749.1 | 17 | 2624 | |||
XM_008769528.1 | XP_008767750.1 | 17 | 2624 | |||
XM_008769529.1 | XP_008767751.1 | 17 | 2624 | |||
XM_008769530.1 | XP_008767752.1 | 16 | 3222 | |||
GenBank mRNA | AY057064.1 | - | 14 | 1653 |