Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130746.1 | NP_570102.1 | 10 | 1352 | ||
GenBank mRNA | AF026554.1 | - | 10 | 1352 | ||
AF081204.2 | - | 7 | 940 | |||
AF081205.1 | - | 7 | 940 | |||
BC168145.1 | - | 8 | 1214 | 8371761 |