Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tob1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133317.2 | NP_579851.2 | 1 | 1178 | ||
GenBank mRNA | AF349723.1 | - | 1 | 1170 |