Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034830.1 | NP_001030002.1 | 7 | 908 | ||
NM_001034831.1 | NP_001030003.1 | 7 | 907 | |||
NM_133429.2 | NP_596920.2 | 7 | 991 | |||
XM_006237326.2 | XP_006237388.1 | 5 | 769 | |||
XM_006237327.2 | XP_006237389.1 | 8 | 956 | |||
XM_006237328.2 | XP_006237390.1 | 5 | 704 | |||
XM_006237329.2 | XP_006237391.1 | 6 | 817 | |||
GenBank mRNA | AB019281.1 | - | 7 | 908 | ||
AF216804.1 | - | 7 | 991 | |||
AF216805.1 | - | 5 | 710 | |||
AF216806.1 | - | 7 | 907 | |||
AF216807.1 | - | 8 | 881 |