Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133562.1 | NP_598246.1 | 6 | 1764 | ||
XM_006251155.2 | XP_006251217.1 | 6 | 1785 | |||
XM_008770248.1 | XP_008768470.1 | 6 | 1806 | |||
GenBank mRNA | AB032551.1 | - | 6 | 1764 |