Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133580.1 | NP_598264.1 | 5 | 320 | ||
GenBank mRNA | BC061984.1 | - | 6 | 628 | 5599180 | |
U18771.1 | - | 5 | 320 |