Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133590.1 | NP_598274.1 | 2 | 494 | ||
XM_006249744.2 | XP_006249806.1 | 3 | 471 | |||
XM_006249745.2 | XP_006249807.1 | 3 | 328 | |||
XM_008769422.1 | XP_008767644.1 | 1 | 29 | |||
GenBank mRNA | BC081732.1 | - | 3 | 450 | 7109012 | |
FQ233577.1 | - | 2 | 260 | |||
FQ234855.1 | - | 2 | 260 | |||
X96663.1 | - | 2 | 494 |