Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276429.1 | NP_001263358.1 | 4 | 286 | ||
NM_001276430.1 | NP_001263359.1 | 3 | 271 | |||
NM_133604.1 | NP_598288.1 | 4 | 357 | |||
GenBank mRNA | CB775939.1 | - | 4 | 336 | ||
FM052814.1 | - | 4 | 195 | |||
FM056491.1 | - | 3 | 234 | |||
FQ228524.1 | - | 4 | 357 |