Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 2 | 134 | ||
GenBank mRNA | BC063150.1 | - | 2 | 132 | 6922267 | |
BP494884.1 | - | 2 | 133 | |||
CB711033.1 | - | 2 | 150 | |||
FJ179404.1 | - | 1 | 63 | |||
FQ209464.1 | - | 2 | 147 | |||
FQ209465.1 | - | 2 | 154 | |||
FQ211049.1 | - | 2 | 154 | |||
FQ212841.1 | - | 2 | 155 | |||
FQ218096.1 | - | 2 | 137 | |||
FQ218830.1 | - | 2 | 162 | |||
FQ219010.1 | - | 2 | 156 | |||
FQ219410.1 | - | 2 | 128 | |||
FQ219464.1 | - | 2 | 160 | |||
FQ219581.1 | - | 2 | 156 | |||
FQ219850.1 | - | 2 | 146 | |||
J03752.1 | - | 2 | 118 |