Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175838.1 | NP_787032.1 | 4 | 643 | ||
GenBank mRNA | BC063162.1 | - | 4 | 639 | 6918979 | |
BC072542.1 | - | 4 | 638 | 7103726 | ||
BC091297.1 | - | 4 | 624 | 7309009 | ||
BC111707.1 | - | 3 | 607 | 7367097 | ||
BC128723.1 | - | 4 | 618 | 7312569 | ||
X61043.1 | - | 4 | 643 | |||
X63561.1 | - | 3 | 616 |