Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134375.3 | NP_599202.2 | 2596 | |||
GenBank mRNA | DQ631800.1 | - | 2596 | |||
M85183.1 | - | 1472 |
Gene Symbol: LOC103690037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230507.2 | XP_006230569.2 | 7 | 3760 |