Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134376.1 | NP_599203.1 | 4 | 677 | ||
GenBank mRNA | AJ431642.1 | - | 4 | 677 |