Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007149.1 | NP_001007150.1 | 9 | 1578 | ||
NM_001007150.1 | NP_001007151.1 | 7 | 1305 | |||
NM_134466.2 | NP_604461.2 | 8 | 1401 | |||
GenBank mRNA | AF483620.1 | - | 9 | 1578 | ||
AF483621.1 | - | 9 | 1578 | |||
AY549445.1 | - | 7 | 1305 | |||
AY549446.1 | - | 7 | 1305 | |||
AY549447.1 | - | 8 | 1401 | |||
AY549448.1 | - | 8 | 1401 | |||
AY684789.1 | - | 1305 | ||||
AY684790.1 | - | 1305 | ||||
BC085705.1 | - | 1937 | 7190916 |