Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahnak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191951.1 | NP_001178880.1 | 6 | 12153 | ||
XM_008760214.1 | XP_008758436.1 | 5 | 12554 |