Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138845.2 | NP_620200.2 | 10, 11 | 3554 | ||
XM_006242061.2 | XP_006242123.1 | 6, 7 | 3663 | |||
XM_006242063.2 | XP_006242125.1 | 6, 7 | 3662 | |||
XM_006242064.2 | XP_006242126.1 | 5, 6 | 3503 | |||
GenBank mRNA | AF275151.1 | - | 2, 3 | 316 |