Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138846.1 | NP_620201.1 | 7 | 972 | ||
XM_006256850.2 | XP_006256912.1 | 8 | 1513 | |||
GenBank mRNA | AB036421.1 | - | 7 | 972 |