Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138890.2 | NP_620245.2 | 1, 2 | 624 | ||
GenBank mRNA | AF494093.2 | - | 1, 2 | 624 |