Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 3 | 236 | ||
NM_001271536.1 | NP_001258465.1 | 3 | 322 | |||
NM_012495.2 | NP_036627.1 | 3 | 338 | |||
XM_006230209.2 | XP_006230271.1 | 5 | 584 | |||
XM_006230210.2 | XP_006230272.1 | 4 | 494 | |||
XM_006230211.1 | XP_006230273.1 | 4 | 349 | |||
GenBank mRNA | BC064440.1 | - | 3 | 315 | 5623628 | |
CB750468.1 | - | 3 | 322 | |||
CF975930.1 | - | 3 | 296 | |||
CO398643.1 | - | 3 | 229 | |||
FQ212782.1 | - | 3 | 289 | |||
FQ213869.1 | - | 3 | 319 | |||
FQ214123.1 | - | 3 | 289 | |||
FQ214510.1 | - | 3 | 291 | |||
FQ216189.1 | - | 3 | 244 | |||
FQ216820.1 | - | 3 | 240 | |||
FQ224221.1 | - | 3 | 329 | |||
FQ228954.1 | - | 3 | 268 | |||
FQ229100.1 | - | 3 | 283 | |||
M12919.1 | - | 3 | 338 | |||
M14420.1 | - | 3 | 236 |