Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177305.1 | NP_001170776.1 | 3 | 318 | ||
NM_001271536.1 | NP_001258465.1 | 3 | 404 | |||
NM_012495.2 | NP_036627.1 | 3 | 420 | |||
XM_006230209.2 | XP_006230271.1 | 5 | 666 | |||
XM_006230210.2 | XP_006230272.1 | 4 | 576 | |||
XM_006230211.1 | XP_006230273.1 | 4 | 431 | |||
GenBank mRNA | BC064440.1 | - | 3 | 397 | 5623628 | |
CO398643.1 | - | 3 | 311 | |||
FQ212782.1 | - | 3 | 371 | |||
FQ213869.1 | - | 3 | 401 | |||
FQ214123.1 | - | 3 | 371 | |||
FQ214510.1 | - | 3 | 373 | |||
FQ216189.1 | - | 3 | 326 | |||
FQ216820.1 | - | 3 | 322 | |||
FQ224221.1 | - | 3 | 411 | |||
FQ228954.1 | - | 3 | 350 | |||
FQ229100.1 | - | 3 | 365 | |||
M12919.1 | - | 3 | 420 | |||
M14420.1 | - | 3 | 318 |